Order Entry
United States
Orders LinkContactUsLinkComponent
M-<i>Enterococcus</i> Agar Base Dehydrated Culture Media, HiMedia

M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia

M-Enterococcus Agar Base
Catalog # 95020-490
Supplier:   HiMedia
Supplier #:  M1108-500G
M-<i>Enterococcus</i> Agar Base Dehydrated Culture Media, HiMedia
Catalog # 95020-490
Supplier:  HiMedia
CAS Number:  
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia

M-Enterococcus Agar Base

CAS Number:  
Price
$203.00
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

For the isolation and enumeration of enterococci in water, sewage, food and other materials by membrane filter technique as well as direct plating of specimens.

  • Microbiological culture media
  • 500 g poly bottles
Specifications

Specifications

  • Description:
    M-Enterococcus Agar Base

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.