Orders LinkContactUsLinkComponent
BD GasPak™ EZ Container Systems, BD Diagnostics
BD GasPak™ EZ Container Systems, BD Diagnostics
Incubation Chamber, Large
Catalog # 90003-636
Supplier:   BD
Supplier #:  260672
BD GasPak™ EZ Container Systems, BD Diagnostics
Catalog # 90003-636
Supplier:  BD
BD GasPak™ EZ Container Systems, BD Diagnostics
BD GasPak™ EZ Container Systems, BD Diagnostics
Incubation Chamber, Large
Price
$284.13
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rapid gas generating sachets produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments.
Pouch systems include 20 resealable pouches and 20 rapid gas-generating sachets. The Anaerobe Pouch System also includes 20 dry anaerobic indicators. Sachets do not require water or a catalyst to produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments. Pouches are one-step resealable; no sealing bars needed. Multi-layer plastic pouch material preserves environmental integrity for maximum performance. Pouches can accommodate up to four Petri dishes.
Sachets do not require water or a catalyst. Two incubation container sizes are available: standard and large. These containers are nonbreakable and chemical-resistant for maximum safety. Easy-close latches assure a secure, airtight seal. Containers are designed to hold either petri dish racks or standard test tube racks for greater convenience. Standard container and rack holds 15 to 18 petri dishes and the large container and rack holds 30 to 33 petri dishes.
Ordering information: When using the anaerobic system, an anaerobic indicator must be used. Dry indicator strips change from blue to colorless in the absence of oxygen and may be stored at room temperature prior to use.
Specifications
  • Description:
    Incubation Chamber, Large
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Anaerobic Indicator Strips, Dry, Pack of 100
    Anaerobic Indicator Strips, Dry, Pack of 100
    90003-866
    Log in for availability
    Pack of 100 - $296.01