Order Entry
United States
Orders LinkContactUsLinkComponent
Inoculum Broth, BD

Inoculum Broth, BD

Difco™ Micro Inoculum Broth, 500 g
Catalog # 90000-366
Supplier:   BD
Supplier #:  211813
Inoculum Broth, BD
Catalog # 90000-366
Supplier:  BD
CAS Number:  
Inoculum Broth, BD

Difco™ Micro Inoculum Broth, 500 g

CAS Number:  
Price
$552.42
New Customer Promotional Price
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.

  • Used for cultivating lactobacilli
  • For preparing the inoculum for microbiological assays
  • Peptone provides nitrogen and amino acids
  • Beige, homogenous appearance when dehydrated

Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.

Specifications

Specifications

  • Description:
    Difco™ Micro Inoculum Broth
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.