Orders LinkContactUsLinkComponent
MICROMESH™ Biopsy Processing and Embedding Cassettes, Simport Scientific
MICROMESH™ Biopsy Processing and Embedding Cassettes, Simport Scientific
White
Catalog # 87002-566
Supplier:   Simport Scientific
Supplier #:  M507-2
MICROMESH™ Biopsy Processing and Embedding Cassettes, Simport Scientific
Catalog # 87002-566
Supplier:  Simport Scientific
MICROMESH™ Biopsy Processing and Embedding Cassettes, Simport Scientific
MICROMESH™ Biopsy Processing and Embedding Cassettes, Simport Scientific
White
Price
$1,054.20
Log in for availability
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cassettes with lids are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The one-piece integral lid eliminates the need for separate steel lids. It can be opened and closed as often as necessary and will relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45° angle, making the cassette more compatible with automated cassette printers.
Cassettes are totally resistant to the chemical action of histological solvents. Cassettes have 0.38mm square openings, allowing for maximum fluid exchange without having to use biopsy pads. Large anterior and posterior slots in both the cassette and the cover ensure that the cassette will sink rapidly. Cassettes are 6mm high and feature a 27mm square compartment for needle biopsies. The cover does not protrude above the cassette, allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
Ordering information: Available in 11 noncytotoxic, nonmetallic colors and three fluorescent colors. Each case contains four dispenser boxes of 250 cassettes with the lids closed.
Specifications
  • Color:
    White