Retic-Chex® Linearity Controls, Streck
Retic-Chex® Linearity Controls, 2.5 mL, Plastic Pierceable Vials
Catalog # 82030-690
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Controls are designed to establish the patient reportable range and the analytic linearity of the system
Retic-Chex stain is buffered and filtered new methylene blue that can be used with control and patient samples. It is packaged in a 3 mL plastic squeeze dropper vial and can be stored at room temperature.
Available in five-vial sets with values assigned. Open-vial stability is five days and closed-vial stability is 75 days. Compatible with Abbott®, Beckman Coulter®, and Sysmex® instruments.
Ordering information: Each control is supplied with five vials, one each for levels 1, 2, 3, 4 and 5. The 3.0 mL control is designed specifically for Beckman Coulter® LH 750/LH 755 instruments.
Specifications
Specifications
- Pack type:Plastic Pierceable Vials
- Description:Retic-Chex® Linearity Controls
- Size:2.5 mL



