Order Entry
United States
Orders LinkContactUsLinkComponent
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Green
  25608-854
Supplier #:  4183
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura
  25608-854
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura
Tissue-Tek® Processing/Embedding Cassette Systems, Sakura

Green

 
$296.00
   
Case of 1,500
 
   
 

Cassette systems (cassette, cover, and base) are designed for processing, embedding, sectioning, and storage

Reusable base molds enable perfect specimen positioning. Extended fins prevent tipping and provide gripping surface. Stainless steel construction ensures uniform heating and cooling.

Note: Clear cassettes are polypropylene and are not recommended for use in processors using heat or vacuum.

Plastic cassettes feature an angled, textured writing surface. Available in clear and in 10 colors.

 : Reusable stainless steel cover sold separately.

 
  • Color:
    Green
Accessories

 

  •  
    Description
    Length
    Width
     
     
  • 25608-826
    Base Mold
    15 mm
    15 mm
       
    Case of 12 - $116.00

 

  •  
    Description
    Length
    Width
     
     
  • 25608-832
    Base Mold
    37 mm
    24 mm
       
    Case of 12 - $116.00

 

  •  
    Description
    Length
    Width
     
     
  • 25608-830
    Base Mold
    30 mm
    24 mm
       
    Case of 12 - $116.00

 

  •  
    Description
    Length
    Width
     
     
  • 25608-824
    Base Mold
    7 mm
    7 mm
       
    Case of 12 - $116.00

 

  •  
    Description
     
     
  • 25608-870
    Stainless Steel Cover
       
    Case of 25 - $70.50

 

  •  
    Description
    Length
    Width
     
     
  • 25608-828
    Base Mold
    24 mm
    24 mm
       
    Case of 12 - $116.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.