Trypsin Protease, HyClone™
Trypsin, 0.05% 1X, with 0.5 g porcine trypsin in HBSS with 0.2 g/L EDTA, without calcium, magnesium (liquid), 100 ml
Supplier: Cytiva
: SH30236.01
Trypsin Protease, HyClone™
Catalog # 16777-202
Supplier: Cytiva
Product available on GSA Advantage®
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
Specifications
Specifications
- Description:Trypsin, 0.05% 1X, with 0.5 g porcine trypsin in HBSS with 0.2 g/L EDTA, without calcium, magnesium (liquid)
- Size:100 ml
- Storage Conditions:Less than 0 °C
- Cat. No.:16777-202
- Formulation:Trypsin, 0.05% 1X Solution, with 0.5 g Porcine Trypsin (1:250/L) in HBSS with 0.2 g/L EDTA, without Calcium or Magnesium
- Species:Pig
- Enzyme Name:Trypsin



