Order Entry
United States
Orders LinkContactUsLinkComponent
 

cis-Stilbene ≥96.0%

5 ml
Catalog # TCS0089-005ML
Supplier:   TCI America
 :  S0089-5ML
cis-Stilbene ≥96.0%
Catalog # TCS0089-005ML
Supplier:  TCI America
CAS Number:  645-49-8
cis-Stilbene ≥96.0%
cis-Stilbene ≥96.0%

5 ml

CAS Number:  645-49-8
Price
$82.00
Log in for availability
Each (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 645-49-8 MDL Number: MFCD00004788 Molecular Formula: C14H12 Molecular Weight: 180.25 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Color: Very Pale Yellow Boiling point (°C): 152 Flash Point (°C): 110 Specific Gravity (20/20): 1.02

Other Product Names / Synonyms: cis-1,2-Diphenylethylene

Synonyms:  cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene
  • CAS Number:645-49-8
  • Formula:(C₆H₅)CHCH(C₆H₅)
  • MDL Number:MFCD00004788
  • Molecular Weight:180.25 g/mol
  • Merck Index:12,08972
  • Boiling Pt:152 °C
  • Density:1.02 (20 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:211-445-7
Specifications

Specification Test Results

  • Purity/Analysis Method:
    96.0% (GC)

Specifications

  • Size:
    5 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.