Order Entry
United States
Orders LinkContactUsLinkComponent
Boron trifluoride diethyl ether complex ≥98%

Boron trifluoride diethyl ether complex ≥98%

500 ml
Catalog # TCB0527-500ML
Flammable
Toxic
Corrosive
Health
Supplier:   TCI America
Supplier #:  B0527-500ML
Boron trifluoride diethyl ether complex ≥98%
Catalog # TCB0527-500ML
Supplier:  TCI America
CAS Number:  109-63-7
Boron trifluoride diethyl ether complex ≥98%

500 ml

CAS Number:  109-63-7
Price
$92.40
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 109-63-7 Molecular Formula: C4H10BF3O Molecular Weight: 141.93 Form: Liquid Color: Colorless

Product Categories: Analytical Science, NMR Spectrometry, Bioscience, Sugars, Synthetic Organic Chemistry, Carbonyl Olefination, Functional Group Transformation and Introducing Reaction, Multicomponent Reaction (MCR)

Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:13,01340
  • Boiling Pt:126 °C (1013 hPa)
  • Melting Pt:–60 °C
  • Density:1.125 g/cm³ (25 °C)
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity(Gravimetric Method):
    98.0 to 102.0 %
  • Specific gravity (20/20):
    1.1250 to 1.1320
  • Refractive index n20/D:
    1.3430 to 1.3470

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.