QuEChERS SPE Tubes, Restek
Graphite Carbon, 1000 mg, 500 mg Graphitized Carbon and 500 mg PSA
Catalog # RK26129
Supplier: Restek
Supplier #: 26129
Jump to:
Product Details & Documents
These tubes comply with the USDA's QuEChERS method, used in the extraction and clean-up of pesticide residue samples.
- Contained bed weight varies between models
- Adsorbants and secondary amine exchange materials differ between models
Tubes contain ultrapure adsorbants and primary and secondary amine exchange material (PSA). Fluoropolymer resin frits minimize background.
Specifications
- Bed weight:1000 mg
- Packing material:500 mg Graphitized Carbon and 500 mg PSA
- Phase:Graphite Carbon