

2-Propanol ā„99.9% (by GC), OmniSolvĀ®, SupelcoĀ®
Catalog # EM-PX1834-1
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Cat. No.:EM-PX1834-1
- Supplier no.:PX1834-1
Specifications
Specification Test Results
- Assay (GC):99.9% min
- Capillary ECD responsive substances (as PCNB):2ng/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):250ppt max
- Form:Clear liquid
- Identity (IR-spectrum):Conforms
- Residue after evaporation:1ppm max
- Titrable acid:0.2µeq/g max
- UV Abs. at 204 nm:1.00AU max
- UV Abs. at 205 nm:0.80AU max
- UV Abs. at 210 nm:0.35AU max
- UV Abs. at 220 nm:0.10AU max
- UV Abs. at 230 nm:0.05AU max
- UV Abs. at 240 nm:0.02AU max
- UV Abs. at 260 nm:0.005AU max
- UV Abs. at 300 nm:0.005AU max
- UV Cut-off:204nm max
- Water (H2O):0.05% max
Specification Test Results
About this item
CAS Number:67-63-0
:
Formula:(CHā)āCHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (lit.)
Melting Pt:ā89.5 °C (lit.)
Density:0.785 g/ml (25°C) (lit.)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
Frequently Bought Together