Order Entry
United States
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB) 98%

Bottle, 5 g
Catalog # AAA13868-06
Irritant
Health
 :  A13868-06
3,3',5,5'-Tetramethylbenzidine (TMB) 98%
Catalog # AAA13868-06
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) 98%
3,3',5,5'-Tetramethylbenzidine (TMB) 98%

Bottle, 5 g

CAS Number:  54827-17-7
Price
$189.31
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood.

It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:168…171 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.