Order Entry
United States
Orders LinkContactUsLinkComponent
 

n-Decane 99+%

Bottle, 10 g
Catalog # AA31209-09
Health
Flammable
 :  031209-09
n-Decane 99+%
Catalog # AA31209-09
CAS Number:  124-18-5
n-Decane 99+%
n-Decane 99+%

Bottle, 10 g

CAS Number:  124-18-5
Price
$49.44
Log in for availability
Each (10g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00008954 Beilstein Registry No.: 1696981

Purity: 99+% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colorless Odor: Petrol-like Melting Point: -30° Boiling Point: 172-174° Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954

  • CAS Number:124-18-5
  • Formula:H₃C(CH₂)₈CH₃
  • MDL Number:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:172…174 °C
  • Melting Pt:–30 °C
  • Density:0.730
  • Flash Pt:46 °C (115 °F)
  • Storage Temperature:Ambient
  • EINECS:204-686-4
  • UN:2247
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    10 g
  • Cat. No.:
    AA31209-09

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.