Orders LinkContactUsLinkComponent
VWR® Blocking Buffers
VWR® Blocking Buffers
PBS Containing 3% Non-Fat Powdered Milk, 5 pack
Catalog # 97063-986
Supplier #:  M232-39.8G-5PK
VWR® Blocking Buffers
Catalog # 97063-986
Supplier:  
CAS Number:  
VWR® Blocking Buffers
VWR® Blocking Buffers
PBS Containing 3% Non-Fat Powdered Milk, 5 pack
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Most routinely used blocking buffers in a variety of formats for convenience.
  • Liquid or pre-measured powder Ready-Packs™ available for easy use
  • Optimize assay performance using high quality blocking buffers
Each pack of powdered blocking buffer makes one liter of 1X solution.
Specifications
Specifications
  • Description:
    PBS Containing 3% Non-Fat Powdered Milk
  • Size:
    5 pack