Order Entry
United States
Orders LinkContactUsLinkComponent
Trypsin Protease, HyClone™

Trypsin Protease, HyClone™

Trypsin 2.5% 10X solution, with 25 g porcine trypsin in HBSS, without calcium, magnesium, EDTA or phenol red (liquid), 100 ml
Catalog # 95053-260
Supplier:   Cytiva
Supplier #:  SV30037.01
Trypsin Protease, HyClone™
Catalog # 95053-260
Supplier:  Cytiva
CAS Number:  
Trypsin Protease, HyClone™
Trypsin Protease, HyClone™

Trypsin 2.5% 10X solution, with 25 g porcine trypsin in HBSS, without calcium, magnesium, EDTA or phenol red (liquid), 100 ml

CAS Number:  
Price
$53.66
Log in for availability
Each (100ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.

  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Specifications

Specifications

  • Description:
    Trypsin 2.5% 10X solution, with 25 g porcine trypsin in HBSS, without calcium, magnesium, EDTA or phenol red (liquid)
  • Size:
    100 ml
  • Storage Conditions:
    Less than 0 °C
  • Cat. No.:
    95053-260
  • Formulation:
    Trypsin, 2.5% 10X Solution, with 25.0 g Porcine Trypsin (1:250/L) in HBSS, without Calcium and Magnesium, without EDTA or Phenol Red
  • Species:
    Pig
  • Enzyme Name:
    Trypsin

Specifications