Order Entry
United States
Orders LinkContactUsLinkComponent
BBL™ Phosphate Buffer, BD

BBL™ Phosphate Buffer, BD

Phosphate buffer, 500 ml, 6.8
Catalog # 90004-948
Supplier:   BD
Supplier #:  297657
BBL™ Phosphate Buffer, BD
Catalog # 90004-948
Supplier:  BD
CAS Number:  
BBL™ Phosphate Buffer, BD

Phosphate buffer, 500 ml, 6.8

CAS Number:  
Price
$108.90
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This is used as buffered diluent for microbiological procedures.

Specifications

Specifications

  • Description:
    Phosphate buffer
  • Size:
    500 ml
  • pH:
    6.8
  • Cat. No.:
    90004-948
  • Shelf Life:
    365

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.