Orders LinkContactUsLinkComponent
Difco™ m Enterococcus Agar, BD Diagnostics
Difco™ m Enterococcus Agar, BD Diagnostics
Catalog # 90003-930
Difco™ m Enterococcus Agar, BD Diagnostics
Catalog # 90003-930
Supplier:  
CAS Number:  
Difco™ m Enterococcus Agar, BD Diagnostics
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MEMBRANE FILT ENTEROCOCCI 500G Catalog #EM1.05289.0500
Product EM1.05289.0500 is a replacement for 90003-930 but varies by Brand, Packaging or Quantity.
MEDIUM M-ENTEROCOCCUS AGAR BASE 500G Catalog #95020-490
Product 95020-490 is a replacement for 90003-930 but varies by Brand, Packaging or Quantity.
M-ENTERCCCUS HVEG AGARBASE500G Catalog #61000-458
Product 61000-458 is a replacement for 90003-930 but varies by Brand, Packaging or Quantity.
See more alternatives
Product Details & Documents
Selective isolation and enumeration of enterococci using the membrane filtration technique.
Documents