Orders LinkContactUsLinkComponent
LPM Agar Base, BD™
LPM Agar Base, 500 g
Catalog # 90001-604
LPM Agar Base, BD™
Catalog # 90001-604
Supplier:  
CAS Number:  
LPM Agar Base, BD™
LPM Agar Base, 500 g
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
LPM AGAR BASE 2KG Catalog #90001-602
LPM HIVEG AGAR BASE 100GM Catalog #61000-566
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Use with Moxalactam Antimicrobic supplement for isolation of L
monocytogenes.
Specifications
Specifications
  • Description:
    LPM Agar Base
  • Size:
    500 g
Specifications