Order Entry
United States
Orders LinkContactUsLinkComponent
Difco™ m-EI Modified Agar, BD Diagnostics

Difco™ m-EI Modified Agar, BD Diagnostics

m EI Agar, BD Difco, 500 g
Catalog # 90000-790
Supplier:   BD
Supplier #:  214881
Difco™ m-EI Modified Agar, BD Diagnostics
Catalog # 90000-790
Supplier:  BD
CAS Number:  
Difco™ m-EI Modified Agar, BD Diagnostics

m EI Agar, BD Difco, 500 g

CAS Number:  
Price
$2,504.70
New Customer Promotional Price
Log in for availability
Each (500g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the chromogenic detection and enumeration of Enterococci in water by the membrane filter technique

Conforms with US EPA Approved Methods

Specifications

Specifications

  • Description:
    m EI Agar, BD Difco
  • Size:
    500 g

Specifications