Orders LinkContactUsLinkComponent
Papanicolaou's solution EA 36
Papanicolaou's solution EA 36
473 ml (16 oz.)
Catalog # 84000-114
Supplier:   Epredia
Supplier #:  75404
Papanicolaou's solution EA 36
Catalog # 84000-114
Supplier:  Epredia
CAS Number:  
Papanicolaou's solution EA 36
Papanicolaou's solution EA 36
473 ml (16 oz.)
CAS Number:  
Price
$436.76
New Customer Promotional Price
Log in for availability
Case of 4 (0.25 .pt US)
Product Details & Documents
Control the color hues and intensities of your work with Epredia™ Papanicolaou EA Stain, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Epredia™ EA 36 is used in conjunction with Epredia™ OG 6 for the staining of gynecological specimens. It will provide a slightly lighter cytoplasmic hue than EA 50. EA 36 is recommended for end users who desire a more pastel staining result in gynecological specimens.
Documents
Specifications
Specifications
  • Size:
    473 ml (16 oz.)