Order Entry
United States
Orders LinkContactUsLinkComponent
CD-Chex CD34™ Flow Cytometry Controls, Streck

CD-Chex CD34™ Flow Cytometry Controls, Streck

Level 3, 2 x 1.0 mL
Catalog # 82030-694
Supplier:   Streck Laboratories
Supplier #:  213383
CD-Chex CD34™ Flow Cytometry Controls, Streck
Catalog # 82030-694
CAS Number:  
CD-Chex CD34™ Flow Cytometry Controls, Streck
CD-Chex CD34™ Flow Cytometry Controls, Streck

Level 3, 2 x 1.0 mL

CAS Number:  
Price
$534.64
Log in for availability
Pack of 2 (1ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CD-Chex CD34™ is a positive procedural control for enumerating CD34 positive cells by flow cytometry

The CD34 positive cells have characteristics of human hematopoietic progenitor cells. CD-Chex CD34 is available in three levels, providing reference values similar to levels found in normal peripheral blood, bone marrow, cord blood and mobilized patient samples. It is compatible with ISHAGE gating strategies as well as commercially available enumeration kits. CD-Chex CD34 has 90-day closed-vial stability and 30-day open-vial stability.

Specifications

Specifications

  • Description:
    Level 3
  • Size:
    2 x 1.0 mL

Specifications