Order Entry
United States
Orders LinkContactUsLinkComponent
Poly(caprolactone), soluble

Poly(caprolactone), soluble

Glass, amber, 500 g
Catalog # 700012-648
Supplier #:  P1705-500GM
Poly(caprolactone), soluble
Catalog # 700012-648
CAS Number:  24980-41-4
Poly(caprolactone), soluble

Glass, amber, 500 g

CAS Number:  24980-41-4
Price
$840.88
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Poly(caprolactone), Flakes is a biodegradable polymer often used as an additive for resins to improve their processing characteristics and their end use properties (e.g., impact resistance).

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  6-Caprolactone polymer, 2-Oxepanone homopolymer, PCL
  • CAS Number:24980-41-4
  • Formula:(C₆H₁₀O₂)n
  • MDL Number:MFCD00084404
  • Molecular Weight:114.1400 g/mol
  • Density:1.146
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass, amber
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.