Order Entry
United States
Orders LinkContactUsLinkComponent
 

1,2,6-Hexanetriol ≥97% for synthesis

Glass, amber, 25 g
  700003-038
 :  T2117-25GM
1,2,6-Hexanetriol ≥97% for synthesis
  700003-038
 :  106-69-4
1,2,6-Hexanetriol ≥97% for synthesis

Glass, amber, 25 g

 :  106-69-4
 
$112.95
   
Each (25g)
 
   
 

1,2,6-Trihydroxyhexane is a stable, high boiling, liquid polyol that is miscible with water and is non-toxic. 1,2,6-TRIHYDROXYHEXANE is used in cosmetic and pharmaceutical cream formulations as a moisturizing agent (humectant). Its excellent solvent properties also lend it useful in as a carrier solvent for ink and paint formulations, as well as in photographic and textile formulations. Due to the interesting physical and chemical cross-linking properties of this triol, it finds utility in the manufacture of synthetic resins, alkyd resins, plasticizers and polyurethane systems.

 :  1,2,6-Trihydroxyhexane
  •  :106-69-4
  •  :C₆H₁₄O₃
  •  :MFCD00002976
  •  :134.18 g/mol
  •  :323 °C (1013 hPa)
  •  :25…32 °C
  •  :1.109
  •  :Ambient
  •  :203-424-6
 

 

  • Assay:
    97% Min.

 

  • Pack type:
    Glass, amber
  • Size:
    25 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.