Orders LinkContactUsLinkComponent
Attest™ Biological Indicators for EO, 3M™
Attest™ Biological Indicators for EO, 3M™
Biological Indicator‎
Catalog # 56222-516
Supplier:   Healthcare Products
Supplier #:  MMM 1264
Attest™ Biological Indicators for EO, 3M™
Catalog # 56222-516
Attest™ Biological Indicators for EO, 3M™
Biological Indicator‎
Price
$581.72
Log in for availability
Box of 100 Case of 400
Product Details & Documents
3M™ Attest™ self-contained BI is designed for monitoring ethylene oxide (EO) sterilization processes.
It consists of a Bacillus atrophaeus spore strip; sealed glass ampule with growth medium and bromothymol blue pH indicator dye system; green color-coded cap with holes for sterilant penetration and a filter as a bacterial barrier, and a chemical indicator on the label that changes to green when processed. After sterilization, the vial is 'crushed' to join the growth media with the processed spore strip. The BI is incubated for 48 hours for a visual color change readout. A color change to yellow indicates surviving spores and a positive result.
Documents
Specifications
  • Description:
    Biological Indicator‎