Extractor Arms, Nederman
Product Details & Documents
Ideal for applications requiring removal of dust and fumes, these flexible extraction arms feature optimal capturing capacity and maneuverability for easy positioning
Arms are suspended by a robust swivel allowing for 360° rotation and are equipped with a damper in the hood to save energy when not in operation. The durable hood features integrated start/stop switch for fan control and optional lighting. Units can be mounted on brackets, extension arms, exhaust rails, and on fixed or mobile filter equipment. The 2.1m (7') arms are ideal for tight spaces, or where the need for reach is small. The 4.3m (14') arms are available with horizontal or vertical configuration. The horizontal arm has a long reach and is ideal for application in environments with low ceilings. The vertical arm provides long reach for both low and high level extraction.
Ordering information: Fume extractor arms are not supplied with fans. For compatible fans and starters, see 46102-214 series.
Specifications
- Description:Extractor Arm with Blue Hose
- Length:3 m (10')
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- Mounting Bracket
Product Information
- Catalog #ElectricalAvailabilityPrice
- Hood Spotlight
Product Information
- Catalog #AvailabilityPrice
- Spark Screen, Wire Mesh
Product Information
- Catalog #LengthAvailabilityPrice
- Mounting Bracket
Product Information
- Catalog #LengthAvailabilityPrice
- Mounting Bracket