Corning® Minimum Essential Medium (Mod.) (MEM), Corning
MEM, Corning® 50 L MEM (Minimum Essential Medium), powder [+] Earles salts [-] L-glutamine, sodium bicarbonate, Powder
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # 45000-592
CAS Number:
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MEM NEAA 100X SOLN. 100ML PK6 Catalog #45000-700
Product 45000-700 is similar to 45000-592 but manufacturer and specifications may vary.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Documents
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture.pdf Corning Cell Culture Media Quick Reference Guide Updated.pdf Corning Classical Media Formulations H2-2017
Product Literature
More
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Corning Cell Culture Media.pdf
Promotion Material
Specifications
Specifications
- Media Format:Powder
- Description:MEM, Corning® 50 L MEM (Minimum Essential Medium), powder [+] Earles salts [-] L-glutamine, sodium bicarbonate
- Cat. No.:45000-592
- Storage Temperature:Refrigerate



