Orders LinkContactUsLinkComponent
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts [-] L-glutamine, phenol red, sodium bicarbonate, Powder
Catalog # 45000-586
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # 45000-586
Supplier:  
CAS Number:  
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts [-] L-glutamine, phenol red, sodium bicarbonate, Powder
CAS Number:  
Discontinued
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Specifications
Specifications
  • Media Format:
    Powder
  • Description:
    MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts [-] L-glutamine, phenol red, sodium bicarbonate