Orders LinkContactUsLinkComponent
HistoGel™ Specimen Processing Gel, Epredia
HistoGel™ Specimen Processing Gel, Epredia
Cooling Block
Catalog # 16002-560
Supplier:   Epredia
Supplier #:  WB-4010-1
HistoGel™ Specimen Processing Gel, Epredia
Catalog # 16002-560
Supplier:  Epredia
CAS Number:  
HistoGel™ Specimen Processing Gel, Epredia
HistoGel™ Specimen Processing Gel, Epredia
Cooling Block
CAS Number:  
Price
$389.18
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Encapsulate and retain even small, friable and viscous specimens during histological processing with Epredia™ HistoGel™ Specimen Processing Gel.
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
This gel encapsulates and retains specimens during histological processing, regardless of the specimens’ small, friable, or viscous nature. Chemical agents penetrate the specimen without allowing it to escape from the gel. The gel does not retain histological stains, eliminating unwanted discoloration around specimens on slides. It is virtually unnoticeable during sectioning and will not pop out of the paraffin block during cutting.
Ordering information: Starter Kit includes twelve 10ml (0.34oz.) HistoGel™ vials, two WonderBlock cooling blocks, a dry bath incubator, and 500 disposable pipets.
Specifications
Specifications
  • Description:
    Cooling Block