Order Entry
United States
Orders LinkContactUsLinkComponent
 

Glucose Solution, Teknova

Glucose Solution, 50%, Sterile, 1 L
Catalog # 101446-796
Supplier:   Teknova
 :  G0550
Glucose Solution, Teknova
Catalog # 101446-796
Supplier:  Teknova
CAS Number:  
Glucose Solution, Teknova

Glucose Solution, 50%, Sterile, 1 L

CAS Number:  
Price
$164.44
Log in for availability
Each
 
Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Glucose Solution, 50%, Sterile
  • Volume:
    1 L

Specifications