Order Entry
United States
Orders LinkContactUsLinkComponent
 

Glucose Solution, Teknova

Glucose Solution, 20%, Sterile, 500 ml
Catalog # 100216-668
Glucose Solution, Teknova
Catalog # 100216-668
Supplier:  
CAS Number:  
Glucose Solution, Teknova

Glucose Solution, 20%, Sterile, 500 ml

CAS Number:  
Discontinued
Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Glucose Solution, 20%, Sterile
  • Volume:
    500 ml

Specifications