Order Entry
United States
Orders LinkContactUsLinkComponent
Bis(2-ethoxyethyl)ether ≥98.0%

Bis(2-ethoxyethyl)ether ≥98.0%

25 ml
Catalog # TCB0489-025ML
Supplier:   TCI America
Supplier #:  B0489-25ML
Bis(2-ethoxyethyl)ether ≥98.0%
Catalog # TCB0489-025ML
Supplier:  TCI America
CAS Number:  112-36-7
Bis(2-ethoxyethyl)ether ≥98.0%

25 ml

CAS Number:  112-36-7
Price
$19.00
Log in for availability
Each (25ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 112-36-7 MDL Number: MFCD00009254 Molecular Formula: C8H18O3 Molecular Weight: 162.23 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 188 Melting point (°C): -44 Flash Point (°C): 71 Specific Gravity (20/20): 0.91

Product Categories: Materials Chemistry, Resin Additives Other Product Names / Synonyms: Bis(2-ethoxyethyl) Ether, Diethyl Carbitol

Synonyms:  2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether
  • CAS Number:112-36-7
  • Formula:C₈H₁₈O₃
  • MDL Number:MFCD00009254
  • Molecular Weight:162.23 g/mol
  • Merck Index:13,03145
  • Boiling Pt:188 °C
  • Melting Pt:–44 °C
  • Density:0.91 (20 °C)
  • Flash Pt:71 °C
  • Storage Temperature:Ambient
  • EINECS:203-963-7

Documents

Specifications

Specification Test Results

  • Purity/Analysis Method:
    98.0% (GC)

Specifications

  • Size:
    25 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.