Order Entry
United States
Orders LinkContactUsLinkComponent
 

2-(2-Ethoxyethoxy)ethyl acetate 99%

bottle, 2.5 L
Catalog # AAAL13446-0F
Irritant
 :  L13446-0F
2-(2-Ethoxyethoxy)ethyl acetate 99%
Catalog # AAAL13446-0F
CAS Number:  112-15-2
2-(2-Ethoxyethoxy)ethyl acetate 99%

bottle, 2.5 L

CAS Number:  112-15-2
Price
$147.00
Log in for availability
Each (2,500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 112-15-2 Molecular Formula: CH3CO2(CH2CH2O)2C2H5 Formula Weight: 176.21 Flash Point: 95°(203°F) Refractive Index: 1.4210 Melting Point: -25° Boiling Point: 218-219° Beilstein Reference: 1764643 Merck Reference: 14,1800 MDL No.: MFCD00041928

Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Merck Index:14,1800
  • Boiling Pt:218…219 °C
  • Melting Pt:–25 °C
  • Density:1.012
  • Flash Pt:95 °C(203 °F)
  • Storage Temperature:Ambient
  • EINECS:203-940-1
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    2.5 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.