Order Entry
United States
Orders LinkContactUsLinkComponent
1,9-Nonanedithiol 97%

1,9-Nonanedithiol 97%

bottle, 5 g
Catalog # AAAL07172-06
Irritant
Supplier #:  L07172-06
1,9-Nonanedithiol 97%
Catalog # AAAL07172-06
CAS Number:  3489-28-9
1,9-Nonanedithiol 97%

bottle, 5 g

CAS Number:  3489-28-9
Price
$129.27
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 97% CAS No.: 3489-28-9 Molecular Formula: HS(CH2)9SH Formula Weight: 192.39 Flash Point: 100°(212°F) Refractive Index: 1.4999 Boiling Point: 351-352° Beilstein Reference: 1737568 MDL No.: MFCD00004914

Synonyms:  1,9-Dimercaptononane
  • CAS Number:3489-28-9
  • Formula:C₉H₂₀S₂
  • MDL Number:MFCD00004914
  • Molecular Weight:192.39 g/mol
  • Boiling Pt:351…352 °C
  • Melting Pt:–17.5 °C
  • Density:0.953
  • Flash Pt:>100 °C (212 °F)
  • Storage Temperature:Ambient
  • EINECS:222-482-3
  • UN:3334
  • ADR:9,III

Documents

Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.