Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99+%
bottle, 100 g
  AAAA15613-22
 :  A15613-22
L(+)-Ascorbic acid 99+%
  AAAA15613-22
 :  50-81-7
L(+)-Ascorbic acid 99+%
L(+)-Ascorbic acid 99+%
bottle, 100 g
 :  50-81-7
$54.31
Each (100g)
Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328
 :  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  •  :50-81-7
  •  :C₆H₈O₆
  •  :MFCD00064328
  •  :176.13 g/mol
  •  :14,830
  •  :553 °C (1013 hPa)
  •  :190…193 °C (dec.)
  •  :1.65
  •  :Ambient
  •  :200-066-2
  •  :exempt
  • Pack type:
    bottle
  • Size:
    100 g