Order Entry
United States
Orders LinkContactUsLinkComponent
 

Silicon dioxide ≥99.5% (metals basis)

Bottle, 250 g
Catalog # AA89709-30
Health
 :  089709-30
Silicon dioxide ≥99.5% (metals basis)
Catalog # AA89709-30
CAS Number:  7631-86-9
Silicon dioxide ≥99.5% (metals basis)

Bottle, 250 g

CAS Number:  7631-86-9
Price
$70.49
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00011232

Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Synonyms:  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  • CAS Number:7631-86-9
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Merck Index:14,8493
  • Boiling Pt:2230 °C
  • Melting Pt:1710 °C
  • Density:2.2
  • Storage Temperature:Ambient
  • EINECS:231-545-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.