Order Entry
United States
Orders LinkContactUsLinkComponent
pH Indicator solution, wide range pH 4-10

pH Indicator solution, wide range pH 4-10

Dropping bottle, marked, 100 ml
Catalog # 97026-686
Flammable
Irritant
pH Indicator solution, wide range pH 4-10
Catalog # 97026-686
Supplier:  
CAS Number:  
pH Indicator solution, wide range pH 4-10
pH Indicator solution, wide range pH 4-10

Dropping bottle, marked, 100 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
INDICATOR WR4 PH 100ML Catalog #104086-120

Product 104086-120 is similar to 97026-686 but manufacturer and specifications may vary.

PH-INDICATOR SOLUTION (100ML) Catalog #EM1.09175.0100

Product EM1.09175.0100 is similar to 97026-686 but manufacturer and specifications may vary.

Product Details & Documents

For visual determination of pH, using test kits

Method: 10222

  • UN:1219
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Dropping bottle, marked
  • Size:
    100 ml

Specifications