Orders LinkContactUsLinkComponent
Corning® PureCoat™ Flasks, Amine, Corning
Corning® PureCoat™ Flasks, Amine, Corning
75 cm2, 5 Plates/Pack, 5 Packs/Case
Catalog # 95062-844
Supplier:   Corning
Supplier #:  354726
Corning® PureCoat™ Flasks, Amine, Corning
Catalog # 95062-844
Supplier:  Corning
Corning® PureCoat™ Flasks, Amine, Corning
Corning® PureCoat™ Flasks, Amine, Corning
75 cm2, 5 Plates/Pack, 5 Packs/Case
Price
$34.17
New Customer Promotional Price
Log in for availability
Case of 5
Minimum order quantity is 1
Product Details & Documents
PS, with blue screw cap.
Corning® PureCoat™ amine flasks provide an enhanced tissue culture surface. The positively charged surface, manufactured using a proprietary thin-film coating technology, offers improved attachment, increased proliferation, and enhanced recovery from freeze-thaw for a broad range of primary, transfected and fastidious cells that exhibit poor attachment properties on standard tissue culture (TC) surfaces in serum-free/serum-reduced conditions.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
  • Size:
    75 cm2
  • Packed:
    5 Plates/Pack, 5 Packs/Case