Orders LinkContactUsLinkComponent
BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, 10 x 100 mm; 15 x 100 mm
Catalog # 90004-718
Supplier:   BD
Supplier #:  297030
BBL™ Standard Methods Agar, BD Life Sciences
Catalog # 90004-718
Supplier:  BD
CAS Number:  
BBL™ Standard Methods Agar, BD Life Sciences
BBL™ Standard Methods Agar, 10 x 100 mm; 15 x 100 mm
CAS Number:  
Price
$92.37
New Customer Promotional Price
Log in for availability
Pack of 20
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Standard Methods Agar is used for obtaining microbial plate counts from milk and dairy products, foods, water, and other materials of sanitary importance.
  • Enzymatic digest of casein provides nitrogen and amino acids
  • Yeast extract supplies B-complex vitamins
  • Dextrose is an energy source
  • Agar is the solidifying agent
Standard Methods Agar is made according to the American Public Health Association (APHA) formulation.
Specifications
Specifications
  • Description:
    BBL™ Standard Methods Agar
  • Size:
    10 x 100 mm; 15 x 100 mm
Specifications