Order Entry
United States
Orders LinkContactUsLinkComponent
Microbial Content Test Agar, BD Diagnostics

Microbial Content Test Agar, BD Diagnostics

Microbial Content Test Agar, 500 g
Catalog # 90003-500
Supplier:   BD
Supplier #:  255320
Microbial Content Test Agar, BD Diagnostics
Catalog # 90003-500
Supplier:  BD
CAS Number:  
Microbial Content Test Agar, BD Diagnostics
Microbial Content Test Agar, BD Diagnostics

Microbial Content Test Agar, 500 g

CAS Number:  
Price
$234.63
New Customer Promotional Price
Log in for availability
Each (500g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Microbial Content Test Agar is used for the detection of microorganisms on surfaces sanitized with quaternary ammonium compounds.

  • Casein and peptones provide essential nutrients
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent

Microbial Content Test Agar is also known as Tryptic Soy Agar with Lecithin and Polysorbate 80, and is recommended for the Aerobic Plate Count (Microbial Limit Test) for water-miscible cosmetic products containing preservatives.

Specifications

Specifications

  • Description:
    Microbial Content Test Agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.