Orders LinkContactUsLinkComponent
EC Medium, Modified, BD Biosciences
EC Medium, Modified, 500 g
  90002-666
EC Medium, Modified, BD Biosciences
  90002-666
 :  
 :  
EC Medium, Modified, BD Biosciences
EC Medium, Modified, 500 g
 :  
Modified EC medium is used with novobiocin for the selective pre-enrichment of Escherichia coli.
  • Sodium chloride maintains osmotic balance of the medium
  • Novobiocin suppresses the growth of nuisance organisms
Lactose is the carbon source for EC medium, and bile salts mixtures are used to inhibit some Gram-positive cocci and sporeformers.
  • Description:
    EC Medium, Modified
  • Size:
    500 g