Order Entry
United States
Orders LinkContactUsLinkComponent
LPM Agar Base, BD™

LPM Agar Base, BD™

LPM Agar Base, 2 kg
Catalog # 90001-602
Supplier:   BD
Supplier #:  222110
LPM Agar Base, BD™
Catalog # 90001-602
Supplier:  BD
CAS Number:  
LPM Agar Base, BD™

LPM Agar Base, 2 kg

CAS Number:  
Price
$1,475.10
New Customer Promotional Price
Log in for availability
Each (2kg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Use with Moxalactam Antimicrobic supplement for isolation of L

monocytogenes.

Specifications

Specifications

  • Description:
    LPM Agar Base
  • Size:
    2 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.