Order Entry
United States
Orders LinkContactUsLinkComponent
 

Wright-Giemsa stain

1 L (33.8 oz.)
Catalog # 84000-182
Supplier:   Epredia
 :  89013
Wright-Giemsa stain
Catalog # 84000-182
Supplier:  Epredia
CAS Number:  
Wright-Giemsa stain
Wright-Giemsa stain

1 L (33.8 oz.)

CAS Number:  
Price
$252.89
New Customer Promotional Price
Log in for availability
Each (32oz)
Product Details & Documents

Rapidly and conveniently stain blood smears with Epredia™ Wright Giemsa Stain.

Synonyms:  Wright-Giemsa Solution
  • MDL Number:MFCD00132593
Specifications

Specifications

  • Size:
    1 L (33.8 oz.)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.