Orders LinkContactUsLinkComponent
Dehydrant Blends, Epredia
Dehydrant 80%, Poly Bottle, 4 x 1 gal.
  84000-050
 :   Epredia
 :  6401
Dehydrant Blends, Epredia
  84000-050
 :  Epredia
 :  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia
Dehydrant 80%, Poly Bottle, 4 x 1 gal.
 :  
$416.05
Case of 4 (0.25 .US Gal)
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
  • Pack type:
    Poly Bottle
  • Description:
    Dehydrant 80%
  • Size:
    4 x 1 gal.