Orders LinkContactUsLinkComponent
Dehydrant Blends, Epredia
Dehydrant 100%, Poly Bottle, 1 gal.
Catalog # 66004-946
Supplier:   Epredia
 :  6201
Dehydrant Blends, Epredia
Catalog # 66004-946
Supplier:  Epredia
CAS Number:  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia
Dehydrant 100%, Poly Bottle, 1 gal.
CAS Number:  
Price
$517.38
New Customer Promotional Price
Log in for availability
Case of 4 (1/Itm/Bx)
Product Details & Documents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
Specifications
Specifications
  • Pack type:
    Poly Bottle
  • Description:
    Dehydrant 100%
  • Size:
    1 gal.