Order Entry
United States
Orders LinkContactUsLinkComponent
Staticmaster® Static Eliminating Brushes, NRD

Staticmaster® Static Eliminating Brushes, NRD

3” Static Neutralizing Brush, 7.62 cm (3")
Catalog # 58578-040
Supplier:   NRD Staticmaster
Supplier #:  3C500
Staticmaster® Static Eliminating Brushes, NRD
Catalog # 58578-040
Supplier:  NRD Staticmaster
Staticmaster® Static Eliminating Brushes, NRD
Staticmaster® Static Eliminating Brushes, NRD

3” Static Neutralizing Brush, 7.62 cm (3")

Price
$492.00
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

NRD’s static elimination brushes are used in a wide variety of analytical, photographic and medical environments where the elimination of dust on sensitive tools and objects are necessary.

  • Removes dust particles
  • Neutralizes static

The brushes are hand-crafted with natural hair and contain an alpha ionizing strip which neutralizes the static while the soft brushes simultaneously remove dust particles. The neutralization of the surface with the brushes will allow the surface to stay free of dust.

Specifications
  • Description:
    3” Static Neutralizing Brush
  • Brush head width:
    7.62 cm (3")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.