Order Entry
United States
Orders LinkContactUsLinkComponent
Dulbecco's phosphate-buffered saline (DPBS), Mediatech

Dulbecco's phosphate-buffered saline (DPBS), Mediatech

DPBS, Corning® 10 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium, 10 L, +2 °C…+30 °C
Catalog # 45000-438
Supplier:   Corning
Supplier #:  21-031-LX
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Catalog # 45000-438
Supplier:  Corning
CAS Number:  
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Dulbecco's phosphate-buffered saline (DPBS), Mediatech

DPBS, Corning® 10 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium, 10 L, +2 °C…+30 °C

CAS Number:  
Price
$417.00
Log in for availability
Each (10l)
Minimum order quantity is 1
Product Details & Documents

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

Complete Certificate of Analysis available for each production lot, along with full formulation information.

It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.

Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C

Specifications

Specifications

  • Description:
    DPBS, Corning® 10 L DPBS (Dulbeccos Phosphate Buffered Saline), 1X [-] calcium, magnesium
  • Storage temperature:
    +2 °C…+30 °C
  • Volume:
    10 L
  • Cat. No.:
    45000-438
  • Storage Temperature:
    Ambient

Specifications