Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAL GAS SO2 5PPM 58L IN N2
Catalog # 13453-506
Supplier:
Draeger
SDS
Expand image
CAL GAS SO2 5PPM 58L IN N2
Catalog # 13453-506
Supplier:
Draeger
Supplier Number:
4594961
SDS
Related Information
Documentation
SDS
ghs_4594961.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation
Frequently Bought Together
VWR® Nitrile 200 Examination Gloves
Supplier: VWR International
VWR® Powder-Free Nitrile Examination Gloves
Supplier: VWR International
VWR® Premium Thickness Nitrile Gloves
Supplier: VWR International
UltraSense™ Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
STERLING™ Nitrile and STERLING™ Nitrile-XTRA™ Exam Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
Supreno® SE Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Water Resistant Stopwatch, Sper Scientific
Supplier: Sper Scientific