Orders LinkContactUsLinkComponent
Sampling Tubes and Systems, Drager
Sampling Tubes and Systems, Drager
Activated charcoal tubes, Type B/G
Catalog # 13453-468
Supplier:   Draeger
Supplier #:  8101821
Sampling Tubes and Systems, Drager
Catalog # 13453-468
Supplier:  Draeger
Sampling Tubes and Systems, Drager
Sampling Tubes and Systems, Drager
Activated charcoal tubes, Type B/G
Price
$242.96
Log in for availability
Pack of 10
Minimum order quantity is 1
Product Details & Documents
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
Documents
Specifications
  • Description:
    Activated charcoal tubes, Type B/G