Orders LinkContactUsLinkComponent
Interface Cables
Interface Cables
RS-232 cable, DB9m-DB9f fits AB-S, PB-S, SB, PG-S, AX, MX/UMX, 1 m (3.3 ft.)
Catalog # 11276-866
Supplier:   Mettler Toledo
Supplier #:  11101051
Interface Cables
Catalog # 11276-866
Supplier:  Mettler Toledo
Interface Cables
Interface Cables
RS-232 cable, DB9m-DB9f fits AB-S, PB-S, SB, PG-S, AX, MX/UMX, 1 m (3.3 ft.)
Price
$181.00
Log in for availability
Each
Product Details & Documents
Specifications
  • Description:
    RS-232 cable, DB9m-DB9f fits AB-S, PB-S, SB, PG-S, AX, MX/UMX
  • Length:
    1 m (3.3 ft.)