Order Entry
United States
Orders LinkContactUsLinkComponent
Glucose Solution, Teknova

Glucose Solution, Teknova

Glucose Solution, 40%, Sterile, 500 ml
Catalog # 100216-670
Glucose Solution, Teknova
Catalog # 100216-670
Supplier:  
CAS Number:  
Glucose Solution, Teknova

Glucose Solution, 40%, Sterile, 500 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
50% GLUCOSE SOLUTION 1000 ML Catalog #101446-796

Product 101446-796 is a replacement for 100216-670 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Glucose Solution, 40%, Sterile
  • Volume:
    500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.